Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium hydroxide, 75%(as Cd), pure, Thermo Scientific™

Catalog No. AC453880010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453880010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453880010 Supplier Thermo Scientific Chemicals Supplier No. 453880010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21041-95-2
Molecular Formula CdH2O2
Molecular Weight (g/mol) 146.43
MDL Number MFCD00015995
InChI Key PLLZRTNVEXYBNA-UHFFFAOYSA-L
IUPAC Name cadmium(2+) dihydroxide
SMILES [OH-].[OH-].[Cd++]

Specifications

Melting Point 232.0°C
Quantity 1 kg
Assay Percent Range 75%(as Cd)
Merck Index 15,1625
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol in naoh sol, sol in dil acids, in nh4, or nh4cl sol
Formula Weight 146.41
Percent Purity 75% (as Cd)
Grade Pure
Chemical Name or Material Cadmium hydroxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Suspected of causing genetic defects.
May cause cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated e
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Use personal protective equipment as required.
EINECSNumber 244-168-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.