Learn More
Cadmium ICP Standard, Ricca Chemical
SDP
Catalog No. 17014 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
Catalog No. Quantity
1701-4 120 mL
1 options
Catalog No. 1701-4 Supplier Ricca Chemical Company Supplier No. 17014
1mL = 1mg Cd, 1000ppm Cd. (Cd in 5% HNO3) (Class: 8, UN3264, Pkg Grp: III, Corrosive Liquid, Acidic, Inorganic, n.o.s, Nitric Acid)
Specifications
Quantity 120 mL