Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Iodide, Crystal, 99%, Spectrum™ Chemical
SDP

Catalog No. 18609415 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-415 25 g Amber Glass Bottle
18-609-413 500 g Amber Glass Bottle
18-609-414 2.5 kg Amber Glass Bottle
18-609-412 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-609-415 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C103525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.22
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
IUPAC Name cadmium(2+) diiodide
SMILES [Cd++].[I-].[I-]

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 25 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Cadmium Iodide, Crystal
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.