Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Metal Mossy, Innovating Science™
SCI_ED

Catalog No. S000575 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000575 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger: Fatal if inhaled, suspected of causing genetic defects, may cause cancer, suspected of damaging fertility, suspected of damaging the unborn child, causes damage to organs through prolonged or repeated exposure, very toxic to aquatic life with long lasting effects. WARNING! This product can expose you to chemicals including Cadmium and cadmium compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S000575 Supplier Aldon Corporation Supplier No. IS12000
Add to Cart
Add to Cart
Danger: Fatal if inhaled, suspected of causing genetic defects, may cause cancer, suspected of damaging fertility, suspected of damaging the unborn child, causes damage to organs through prolonged or repeated exposure, very toxic to aquatic life with long lasting effects. WARNING! This product can expose you to chemicals including Cadmium and cadmium compounds, which are known to the State of California to cause cancer and reproductive harm.

Chemical Identifiers

Molecular Weight (g/mol) 112.4 g/mol

Specifications

Quantity 100 g
Form Solid
Percent Purity >99.8%
Grade Reagent
Chemical Name or Material Cadmium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.