Learn More
Strem, An Ascensus Company CAS# 10022-68-1. 100g. Cadmium nitrate tetrahydrate, 99% (ACS). MFCD00149626
SDP
Supplier:  Strem, An Ascensus Company 934815100G
Encompass
CAS# 10022-68-1. 100g. Cadmium nitrate tetrahydrate, 99% (ACS). MFCD00149626. Molecular Weight: 236.41 (308.47). Molecular Formula: Cd(NO3)2∙4H2O. Color/form: colorless, deliquescent xtl. Strem# 93-4815. http://www.strem.com/catalog/v/93-4815/
Catalog No. 50-901-15466