Learn More
Cadmium nitrate tetrahydrate, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 132°C |
| Melting Point | 59°C |
| Color | Colorless or White |
| Physical Form | Crystals or Powder |
| Quantity | 1 kg |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | Cd(NO3)2·4H2O |
| Merck Index | 15, 1627 |
| Solubility Information | Solubility in water: 2150g/L (20°C). Other solubilities: soluble in ammonia,practically insoluble in conc. h2so4,soluble in alcohol,acetone and ethyl acetate |
| IUPAC Name | cadmium(2+);dinitrate;tetrahydrate |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Harmful in contact with skin. Fatal if inhaled. May cause cancer. May cause genetic defects. May damage fertility or the unborn child. Causes damage to organs through prolonge |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/p |
| RTECSNumber | EV1850000 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.