Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Oxalate, MP Biomedicals

Catalog No. ICN210252 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210252 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210252 Supplier MP Biomedicals, Inc Supplier No. 0521025205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 814-88-0
Molecular Formula C2H2CdO4
Molecular Weight (g/mol) 202.448
InChI Key OPPBPDNPEOPGLG-UHFFFAOYSA-N
Synonym cadmium oxalate
PubChem CID 92043676
IUPAC Name cadmium;oxalic acid
SMILES C(=O)(C(=O)O)O.[Cd]

Specifications

Quantity 5 g
Formula Weight 200.42
Chemical Name or Material Cadmium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.