Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cadmium oxide

Catalog No. 2028945G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-289-45G 5 g
20-289-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-289-45G Supplier Sigma Aldrich Supplier No. 2028945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cadmium oxide (CdO) is a semiconducting material with high conductivity which isoptically transparent in the electromagnetic spectra. It has a direct bandgap of 2.28 eV and can form a single crystal material by metal organic vapor phase epitaxy (MOVPE) growth.CdO can be used in the synthesis of cadmium selenide quantum dots for potential applications in photocatalysis. It is used as a transparent conducting oxide (TCO) which can further be used in a variety of opto-electronic devices.

Specifications

CAS 1306-19-0
Quantity 5 g
Molecular Formula CdO
Linear Formula CdO
MDL Number MFCD00010921
Molecular Weight (g/mol) 128.41
Percent Purity ≥99.99% trace metals basis
Density 8.15 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.