Learn More
KPM ANALYTICS NORTH AMERICA CORPORATION KPM Analytics Cadmium Reduction Coil, 15 In Length, Low Maintenance, Regenerable
Catalog No. NC1337923
Encompass
The KPM Analytics Cadmium Reduction Coil is a 15 in component specifically designed for effective nitrate to nitrite reduction in various analytical applications. Its extruded cadmium metal construction ensures longevity and low maintenance, while its regenerative capability allows cleaning in-line with water or HCl. This coil enhances laboratory cleanliness and operational ease compared to traditional granulated columns, making it ideal for automated flow analyzers and complex sample analysis.
Catalog No. NC1337923 Supplier KPM ANALYTICS NORTH AMERICA CORPORATION Supplier No. 165030115
The KPM Analytics Cadmium Reduction Coil is a 15 in component specifically designed for effective nitrate to nitrite reduction in various analytical applications. Its extruded cadmium metal construction ensures longevity and low maintenance, while its regenerative capability allows cleaning in-line with water or HCl. This coil enhances laboratory cleanliness and operational ease compared to traditional granulated columns, making it ideal for automated flow analyzers and complex sample analysis.