Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

Catalog No. SC118100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SC118-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SC118-100

missing translation for 'validMainframeMsg'

For atomic absorption. (1mL = 1mg Cd) Solute: Cadmium Metal Solvent: Dilute Nitric Acid

TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 7440-43-9, 7697-37-2, 7732-18-5
Vapor Pressure 14mmHg at 16°C
Density 1.0g/cm³
Physical Form Liquid
Molecular Formula Cd
Concentration 990 to 1010 ppm
MDL Number MFCD00010914
Color Colorless
pH 0.5
Melting Point 0°C
Boiling Point 100°C
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES [Cd]
IUPAC Name cadmium
Molecular Weight (g/mol) 112.41
Quantity 100 mL
PubChem CID 23973
ChEBI CHEBI:22977
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Cadmium, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.