Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Stearate, MP Biomedicals

Catalog No. ICN212739 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212739 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212739 Supplier MP Biomedicals, Inc Supplier No. 0521273910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acts as a lubricant and heat stabilizer in plastics.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2223-93-0
Molecular Formula C36H70CdO4
Molecular Weight (g/mol) 679.366
InChI Key GWOWVOYJLHSRJJ-UHFFFAOYSA-L
Synonym cadmium stearate, cadmium ii stearate, cadmium distearate, octadecanoic acid, cadmium salt, stabilisator scd, stabilizer scd, alaixol 11, cadmium octadecanoate, kadmiumstearat german, cadmium soaps stearate
PubChem CID 16681
IUPAC Name cadmium(2+);octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Cd+2]

Specifications

Quantity 10 g
Formula Weight 679.36
Grade Not Available
Physical Form Powder
Chemical Name or Material Cadmium Stearate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.