Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfate, 99+%, for analysis

Catalog No. AC270565000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270565000 500 g Glass bottle
AC270560025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270565000 Supplier Thermo Scientific Chemicals Supplier No. 270565000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10124-36-4 | CdO4S | 208.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.46
MDL Number MFCD00010923
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
Synonym cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
PubChem CID 24962
ChEBI CHEBI:50292
IUPAC Name cadmium(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Cd+2]

Specifications

Melting Point 1000.0°C
Color White
pH 3.5 to 6.0 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula CdSO4
Merck Index 15, 1632
Solubility Information Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia
Formula Weight 208.46
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Cadmium sulfate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
May damage fertility
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
EINECSNumber 233-331-6
RTECSNumber EV2850000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.