Learn More
Cadmium sulfate, ACS reagent, anhydrous
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1000.0°C |
| Quantity | 500 g |
| Packaging | Glass bottle |
| Linear Formula | CdSO4 |
| Merck Index | 15,1632 |
| Solubility Information | Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia |
| Formula Weight | 208.46 |
| Grade | ACS Reagent |
| Chemical Name or Material | Cadmium sulfate |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes damage to organs through prolonged or repeated exposure. Fatal if inhaled. May cause genetic defects. May cause cancer. Very toxic to aquatic life with long lasting effects. May damage fertility |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath |
| EINECSNumber | 233-331-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.