Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Sulfate, Anhydrous, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18609426 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-426 25 g Amber Glass Bottle
18-609-423 125 g Amber Glass Bottle
18-609-425 500 g Amber Glass Bottle
18-609-424 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-609-426 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C104725GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
IUPAC Name cadmium(2+) sulfate
SMILES [Cd++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 25 g
Percent Purity 99%
Grade ACS/Reagent
Loss on Drying 0.01
Packaging Amber Glass Bottle
Chemical Name or Material Cadmium Sulfate, Anhydrous

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.