Learn More
Hach Company Cadmium TNTplus Vial Test (0.02-0.30 mg/L Cd), 25 Tests
Supplier:  Hach Company TNT852
Encompass
Cadmium TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Cadmium by the Cadion method.Test determines soluble metal. Add Metals Prep Set (TNT890) to determine total cadmium. Add Calcium Separation Set (TNT892) for sample matrices that contain calcium or magnesium in excess of 50 mg/L.
Catalog No. 50-199-8142