Learn More
INDICAL INC IndiSpin QIAcube HT Pathogen Kit
SDP
Catalog No. NC1722060
Encompass_Preferred
The IndiSpin QIAcube HT Pathogen Kit simplifies automated purification of viral RNA/DNA and bacterial DNA from a wide range of animal samples on the QIAcube HT system from QIAGEN (old kit name: cador Pathogen 96 QIAcube HT Kit).¿One universal instrument protocol for highly reliable and rapid extraction of viral RNA/DNA and bacterial DNA¿Fully automated on QIAGEN's high-throughput QIAcube HT platform¿96 samples can be processed in approximately 90 minutes¿Flexibility in multiples of 8 to process between 24 and 96 samples at a time
Catalog No. NC1722060 Supplier INDICAL INC Supplier No. SP54161
The IndiSpin QIAcube HT Pathogen Kit simplifies automated purification of viral RNA/DNA and bacterial DNA from a wide range of animal samples on the QIAcube HT system from QIAGEN (old kit name: cador Pathogen 96 QIAcube HT Kit).¿One universal instrument protocol for highly reliable and rapid extraction of viral RNA/DNA and bacterial DNA¿Fully automated on QIAGEN's high-throughput QIAcube HT platform¿96 samples can be processed in approximately 90 minutes¿Flexibility in multiples of 8 to process between 24 and 96 samples at a time