Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cadrofloxacin 100mg | 153808-85-6 | 100 MG
SDP

Catalog No. 5000229846
Encompass_Preferred

Cadrofloxacin is an orally active fluoroquinolone antibiotic research chemical with activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria. It is supplied as a powder for laboratory research and analytical applications.

  • High purity: 98.2%.
  • Molecular weight: 411.38 g/mol.
  • Chemical formula: C19H20F3N3O4.
  • CAS number: 153808-85-6.
  • Supplied as a powder, stable at -20°C for long-term storage.
  • Available in 100 MG quantity for research use.

Catalog No. 50-002-29846 Supplier Medchemexpress LLC Supplier No. HY116228100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cadrofloxacin is an orally active fluoroquinolone antibiotic research chemical with activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria. It is supplied as a powder for laboratory research and analytical applications.

  • High purity: 98.2%.
  • Molecular weight: 411.38 g/mol.
  • Chemical formula: C19H20F3N3O4.
  • CAS number: 153808-85-6.
  • Supplied as a powder, stable at -20°C for long-term storage.
  • Available in 100 MG quantity for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.