Learn More
Medchemexpress LLC Cadrofloxacin 10mg | 153808-85-6 | 411.38 g·mol^-1 | C19H20F3N3O4 | 10 MG
SDP
Catalog No. 5000229147
Encompass_Preferred
Cadrofloxacin is a fluoroquinolone antibiotic research compound supplied as a white to off-white solid for laboratory use. It is intended for antibacterial and anti-infection research and is provided with supporting documentation for safe handling and characterization.
  • Fluoroquinolone antibiotic used for antibacterial research.
  • High purity suitable for analytical and experimental work.
  • White to off-white solid powder, convenient for dissolution or formulation.
  • Stable when stored frozen as recommended for long-term storage.
  • Available with data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-29147 Supplier Medchemexpress LLC Supplier No. HY11622810MG
Cadrofloxacin is a fluoroquinolone antibiotic research compound supplied as a white to off-white solid for laboratory use. It is intended for antibacterial and anti-infection research and is provided with supporting documentation for safe handling and characterization.
  • Fluoroquinolone antibiotic used for antibacterial research.
  • High purity suitable for analytical and experimental work.
  • White to off-white solid powder, convenient for dissolution or formulation.
  • Stable when stored frozen as recommended for long-term storage.
  • Available with data sheet, certificate of analysis, and safety data sheet.