Learn More
Medchemexpress LLC Cadrofloxacin 25mg | 153808-85-6 | 25 MG
SDP
Catalog No. 5000226626
Encompass_Preferred
Cadrofloxacin is an orally active fluoroquinolone antibiotic supplied for research use only. It exhibits activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria and is provided as a solid powder for laboratory research and assay development.
  • Orally active fluoroquinolone antibacterial agent.
  • Active against aerobic and anaerobic Gram-positive and Gram-negative bacteria.
  • Supplied as a solid powder suitable for research use.
  • Molecular formula C19H20F3N3O4; molecular weight 411.38 g·mol⁻¹.
  • Typical purity approximately 98.2% as reported by the manufacturer.
Catalog No. 50-002-26626 Supplier Medchemexpress LLC Supplier No. HY11622825MG
Cadrofloxacin is an orally active fluoroquinolone antibiotic supplied for research use only. It exhibits activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria and is provided as a solid powder for laboratory research and assay development.
  • Orally active fluoroquinolone antibacterial agent.
  • Active against aerobic and anaerobic Gram-positive and Gram-negative bacteria.
  • Supplied as a solid powder suitable for research use.
  • Molecular formula C19H20F3N3O4; molecular weight 411.38 g·mol⁻¹.
  • Typical purity approximately 98.2% as reported by the manufacturer.