Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cadrofloxacin 50mg | 153808-85-6 | 411.38 g/mol | C19H20F3N3O4 | 50 MG
SDP

Catalog No. 5000226523
Encompass_Preferred

Cadrofloxacin is a fluoroquinolone-class antibacterial compound for laboratory research. It is a small-molecule agent with formula C19H20F3N3O4 and a molecular weight of 411.38 g/mol, supplied at high purity for experimental and analytical use.

  • Fluoroquinolone antibiotic for research applications.
  • Molecular formula C19H20F3N3O4.
  • Molecular weight 411.38 g/mol.
  • Purity >98.2% (HPLC) as stated by supplier.
  • Available in small pack sizes suitable for laboratory use, including 50 MG.
  • Store and handle according to the supplied safety data sheet.

Catalog No. 50-002-26523 Supplier Medchemexpress LLC Supplier No. HY11622850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cadrofloxacin is a fluoroquinolone-class antibacterial compound for laboratory research. It is a small-molecule agent with formula C19H20F3N3O4 and a molecular weight of 411.38 g/mol, supplied at high purity for experimental and analytical use.

  • Fluoroquinolone antibiotic for research applications.
  • Molecular formula C19H20F3N3O4.
  • Molecular weight 411.38 g/mol.
  • Purity >98.2% (HPLC) as stated by supplier.
  • Available in small pack sizes suitable for laboratory use, including 50 MG.
  • Store and handle according to the supplied safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.