Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cadrofloxacin 5mg | 153808-85-6 | 411.38 g·mol⁻1 | C19H20F3N3O4 | 5 MG
SDP

Catalog No. 5000228103
Encompass_Preferred

Cadrofloxacin is a fluoroquinolone antibiotic supplied for research use. It is provided as a solid for experimental assays, with manufacturer guidance for solubilization and storage to preserve activity.

  • Purity 98.21% as reported by the manufacturer.
  • Molecular weight 411.38 g·mol⁻1.
  • Soluble in DMSO at about 10 mg/mL with warming and pH adjustment.
  • Store powder at -20°C for long-term stability; aliquot solutions to avoid freeze-thaw cycles.
  • Suitable for in vitro antibacterial research and assay development.

Catalog No. 50-002-28103 Supplier Medchemexpress LLC Supplier No. HY1162285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cadrofloxacin is a fluoroquinolone antibiotic supplied for research use. It is provided as a solid for experimental assays, with manufacturer guidance for solubilization and storage to preserve activity.

  • Purity 98.21% as reported by the manufacturer.
  • Molecular weight 411.38 g·mol⁻1.
  • Soluble in DMSO at about 10 mg/mL with warming and pH adjustment.
  • Store powder at -20°C for long-term stability; aliquot solutions to avoid freeze-thaw cycles.
  • Suitable for in vitro antibacterial research and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.