Learn More
Bachem Caerulein Ammonium
SDP
Supplier:  Bachem 4030451.0001
Encompass_Preferred
Caerulein Ammonium, Pyr-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2, 17650-98-5, C58H73N13O21S2, Mr 1352.42, 1mg. Ceruletide, Cerulein. This sulfated peptide isolated first from the skin of the Australian frog Litoria caerulea is a more potent secretagogue than its analog CCK-8. Ceruletide is applied in the diagnosis of pancreatic function and, in high dosage, for the induction of pancreatitis in experimental animals.
Catalog No. 50-259-724