Learn More
CPC Scientific Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2 0.5MG
SDP
Supplier:  CPC Scientific CCKS001A
Encompass
SEQUENCE: Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2ONE-LETTER SEQUENCE: Glp-QD-Tyr(SO3H)-TGWMDF-NH2MOLECULAR FORMULA: C58H73N13O21S2MOLECULAR WEIGHT: 1352.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [17650-98-5]SYNONYMS: Ceruletide, CeruleinRESEARCH AREA: GastrointestinalREFERENCES:A. Anastasi et al., Experientia, 23, 699 (1967)SKU(s): CCKS-001A, CCKS-001B, CCKS-001C
Catalog No. 50-210-9106