Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Caerulein, Sulfated, MP Biomedicals™

Catalog No. MP021528605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021528605 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021528605 Supplier MP Biomedicals™ Supplier No. 02152860.5
Add to Cart
Add to Cart

Supplied as a trifluoroacetate salt. Caerulein, Sulfated, MP Biomedicals is an antagonist of endogenous opiods and a gastroregulatory molecule with actions similar to cholecystokinin. It prevents gall bladder pain, renal colic, and the pain of intermitten

Caerulein is used to study cell signaling mediated by NF-kappaB upregulation of proteins, such as intercellular adhesion molecule-1(ICAM-1), and inflammation related molecules, such as NADPH oxidase and Janus kinase signal transducer.

TRUSTED_SUSTAINABILITY

Specifications

CAS 17650-98-5
Color White
Content And Storage -20°C
Format Powder
Molecular Weight (g/mol) 1352.40
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.