Learn More
Medchemexpress LLC Caerulomycin A 100mg | 21802-37-9 | 100 MG
SDP
Catalog No. 5000229402
Encompass_Preferred
Caerulomycin A is a natural-product antifungal and antibiotic research compound (CAS 21802-37-9) that modulates immune signaling by enhancing TGF-β-Smad3 and suppressing interferon-γ-induced STAT1 pathways. It is supplied as a solid for laboratory research with reported purity ≥98% and molecular weight 229.23 g·mol-1.
  • Antifungal and antibiotic activity.
  • Modulates TGF-β-Smad3 and STAT1 signaling.
  • Supplied as milligram-scale solid for research use only.
  • Reported purity ≥98% (HPLC where reported).
  • Soluble in DMSO; store refrigerated or frozen for long-term stability.
Catalog No. 50-002-29402 Supplier Medchemexpress LLC Supplier No. HY114495100MG
Caerulomycin A is a natural-product antifungal and antibiotic research compound (CAS 21802-37-9) that modulates immune signaling by enhancing TGF-β-Smad3 and suppressing interferon-γ-induced STAT1 pathways. It is supplied as a solid for laboratory research with reported purity ≥98% and molecular weight 229.23 g·mol-1.
  • Antifungal and antibiotic activity.
  • Modulates TGF-β-Smad3 and STAT1 signaling.
  • Supplied as milligram-scale solid for research use only.
  • Reported purity ≥98% (HPLC where reported).
  • Soluble in DMSO; store refrigerated or frozen for long-term stability.