Learn More
Medchemexpress LLC HY-114495 1mg Medchemexpress, Caerulomycin A CAS:21802-37-9 Purity:>98%
SDP
Catalog No. 501969365
Encompass_Preferred
Medchemexpress, HY-114495 1mg Caerulomycin A CAS:21802-37-9 Caerulomycin A (Cerulomycin; Caerulomycin), an antifungal compound, induces generation of T cells, enhances TGF-β-Smad3 protein signaling via suppressing interferon-γ-induced STAT1 signaling. Antifungal and antibiotic activity, and used in autoimmune diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9365 Supplier Medchemexpress LLC Supplier No. HY1144951MG
Medchemexpress, HY-114495 1mg Caerulomycin A CAS:21802-37-9 Caerulomycin A (Cerulomycin; Caerulomycin), an antifungal compound, induces generation of T cells, enhances TGF-β-Smad3 protein signaling via suppressing interferon-γ-induced STAT1 signaling. Antifungal and antibiotic activity, and used in autoimmune diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.