Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Caerulomycin A 25mg | 21802-37-9 | 229.24 g/mol | C12H11N3O2 | 25 MG
SDP

Catalog No. 5000229725
Encompass_Preferred

Caerulomycin A is a research-grade antibiotic small molecule with reported antifungal and immunomodulatory activity. It is supplied as a high-purity solid (CAS 21802-37-9) with molecular weight 229.24 g/mol and is commonly dissolved in DMSO for in vitro use or formulated for in vivo dosing.

  • Antibiotic and antifungal activity observed in biological assays.
  • Useful for in vitro and in vivo research applications.
  • High purity: 98.0%.
  • Molecular weight: 229.24 g/mol; formula C12H11N3O2.
  • Soluble in DMSO (150 mg/mL); in vivo formulations documented at ≥2.5 mg/mL.
  • Available in multiple pack sizes, including 25 mg.

Catalog No. 50-002-29725 Supplier Medchemexpress LLC Supplier No. HY11449525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Caerulomycin A is a research-grade antibiotic small molecule with reported antifungal and immunomodulatory activity. It is supplied as a high-purity solid (CAS 21802-37-9) with molecular weight 229.24 g/mol and is commonly dissolved in DMSO for in vitro use or formulated for in vivo dosing.

  • Antibiotic and antifungal activity observed in biological assays.
  • Useful for in vitro and in vivo research applications.
  • High purity: 98.0%.
  • Molecular weight: 229.24 g/mol; formula C12H11N3O2.
  • Soluble in DMSO (150 mg/mL); in vivo formulations documented at ≥2.5 mg/mL.
  • Available in multiple pack sizes, including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.