Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Caerulomycin A 50mg | 21802-37-9 | 229.24 | C12H11N3O2 | 50 MG
SDP

Catalog No. 5000228196
Encompass_Preferred

Caerulomycin A is an antibiotic bioactive compound supplied as a solid for laboratory research use. The product is characterized with analytical documentation and recommended handling instructions; consult the datasheet, COA, and SDS before use.

  • Purity 98.0%.
  • CAS number 21802-37-9.
  • Molecular formula C12H11N3O2.
  • Molecular weight 229.24.
  • Appearance solid, white to off-white.
  • Solubility in DMSO 150 mg/mL.
  • Available in common pack sizes including 50 mg.
  • Supplied for research use with datasheet, COA, and SDS.

Catalog No. 50-002-28196 Supplier Medchemexpress LLC Supplier No. HY11449550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Caerulomycin A is an antibiotic bioactive compound supplied as a solid for laboratory research use. The product is characterized with analytical documentation and recommended handling instructions; consult the datasheet, COA, and SDS before use.

  • Purity 98.0%.
  • CAS number 21802-37-9.
  • Molecular formula C12H11N3O2.
  • Molecular weight 229.24.
  • Appearance solid, white to off-white.
  • Solubility in DMSO 150 mg/mL.
  • Available in common pack sizes including 50 mg.
  • Supplied for research use with datasheet, COA, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.