Learn More
FIREBIRD OPTICS Calcium Fluoride (CaF2) Circular Optical Window- 13x2mm
SDP
Catalog No. NC1953395
Encompass_Preferred
Calcium Fluoride (CaF2) Circular Optical Window- 13x2mm• Wavelength range: 130nm-9.5µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm
Catalog No. NC1953395 Supplier FIREBIRD OPTICS Supplier No. CAF213X2
Calcium Fluoride (CaF2) Circular Optical Window- 13x2mm• Wavelength range: 130nm-9.5µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm