Learn More
Cayman Chemical Caffeic Acid phenylethyl ester, Purity: ≥98%, CAS: 104594-70-9, 50 mg
SDP
Catalog No. 502892757
Encompass_Preferred
A potent and specific inhibitor of NF-kB that exhibits antimitogenic, anticarcinogenic, antiinflammatory, and immunomodulatory properties
Catalog No. 50-289-2757 Supplier Cayman Chemical Supplier No. 7075050 MG
A potent and specific inhibitor of NF-kB that exhibits antimitogenic, anticarcinogenic, antiinflammatory, and immunomodulatory properties