Learn More
Cayman Chemical Caffeic Acid dimethyl ether, Purity: ≥98%, CAS: 2316-26-9, 50 g
SDP
Catalog No. 502942458
Encompass_Preferred
An ester-derivative of dihydrocaffeic acid with antifibrotic activities; inhibits TGF-β1-induced cell growth in LX-2 hepatic stellate cells at 20 µM; prevents TGF-β1-induced increases in the levels of collagen I, ROS, and NOX4 in LX-2 cells at 20 µM; reduces the serum levels of hyaluronic acid, laminin, procollagen type III, and collagen type IV and the liver levels of hydroxyproline in an ethanol- and carbon tetrachloride-induced rat model of hepatic fibrosis at 20 mg/kg per day
Catalog No. 50-294-2458 Supplier Cayman Chemical Supplier No. 3993350 G
An ester-derivative of dihydrocaffeic acid with antifibrotic activities; inhibits TGF-β1-induced cell growth in LX-2 hepatic stellate cells at 20 µM; prevents TGF-β1-induced increases in the levels of collagen I, ROS, and NOX4 in LX-2 cells at 20 µM; reduces the serum levels of hyaluronic acid, laminin, procollagen type III, and collagen type IV and the liver levels of hydroxyproline in an ethanol- and carbon tetrachloride-induced rat model of hepatic fibrosis at 20 mg/kg per day