Learn More
Enzo Life Sciences Cafestol (50mg). CAS: 469-83-0
SDP
Supplier:  Enzo Life Sciences ALX350220M050
Encompass_Preferred
Inducer of glutathione S-transferases. Has chemoprotective activity, reducing the genotoxicity of several carcinogens. Purity: ≥98% (UHPLC). Solubility: Soluble in DMSO, 100% ethanol or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-200-8784