Learn More
Medchemexpress LLC HY-N6257 5mg Medchemexpress, Cafestol CAS:469-83-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN62575MG
Encompass_Preferred
Medchemexpress, HY-N6257 5mg Cafestol CAS:469-83-0 Cafestol, one of the major components of coffee, is a coffee-specific diterpene from. Cafestol is a ERK inhibitor for AP-1-targeted activity against PGE2 production and the mRNA expression of cyclooxygenase (COX)-2 in LPS-activated RAW264.7 cells. Cafestol has strong inhibitory activity on PGE2 production by suppressing the NF-kB activation pathway. Cafestol contributes to its beneficial effects through various biological activities such as chemopreventive, antitumorigenic, hepatoprotective, antioxidative and antiinflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2949