Learn More
Grainger Cafeteria Tray 18 in L Black PK12
Catalog No. 502753223
Encompass_Preferred
The Carlisle 18" L Black Cafeteria Tray is made of high-density polyethylene plastic with textured edges to prevent slipping. It measures 18" long x 14" wide x 1" high and has a weight capacity of 25 lbs. The tray features molded-in handles for easy carrying and is NSF certified. It can withstand temperatures from -20°F to 160°F. The black color provides an attractive look. This durable tray is ideal for cafeterias, buffets, potlucks, and catering.
Catalog No. 50-275-3223 Supplier Grainger Supplier No. 14D349
The Carlisle 18" L Black Cafeteria Tray is made of high-density polyethylene plastic with textured edges to prevent slipping. It measures 18" long x 14" wide x 1" high and has a weight capacity of 25 lbs. The tray features molded-in handles for easy carrying and is NSF certified. It can withstand temperatures from -20°F to 160°F. The black color provides an attractive look. This durable tray is ideal for cafeterias, buffets, potlucks, and catering.