Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeic acid matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013941 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3941 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3941 Supplier MilliporeSigma Supelco Supplier No. 600181G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Caffeic acid is an antioxidant and the major hydroxycinnamic acid present in wine. It contains a catechol group with an α,β-unsaturated carboxylic acid chain with hepatoprotective properties.

TRUSTED_SUSTAINABILITY

Specifications

CAS 331-39-5
Physical Form Powder
Linear Formula (HO)2C6H3 CH=CHCO2 H
MDL Number MFCD00004392
Melting Point 211°C to 213°C
Synonym 3,4-Dihydroxybenzeneacrylic acid; 3,4-Dihydroxycinnamic acid; 3-(3,4-Dihydroxyphenyl)-2-propenoic acid
Type Organic Standard
Quantity 1 g
Formula Weight 180.16
Percent Purity ≥99% (HPLC)
Grade Matrix Substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.