Learn More
Cayman Chemical Caffeic Acid methyl ester, Purity: ≥95%, CAS: 3843-74-1, 50 mg
SDP
Catalog No. 502967391
Encompass_Preferred
An ester of a naturally occurring phenolic compound that demonstrates antihyperglycemic and antidiabetic activity as well as induces anti-inflammatory and antinociceptive effects
Catalog No. 50-296-7391 Supplier Cayman Chemical Supplier No. 1787350 MG
An ester of a naturally occurring phenolic compound that demonstrates antihyperglycemic and antidiabetic activity as well as induces anti-inflammatory and antinociceptive effects