Learn More
TARGETMOL CHEMICALS INC Caffeic Acid Phenethyl Ester
SDP
Catalog No. 502879851
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Caffeic Acid Phenethyl Ester (Phenylethyl Caffeate) (CAPE) inhibits the activation of nuclear transcription factor NF-kappa B and may suppress p70S6K and Akt-driven signaling pathways, with antineoplastic, cytoprotective and immunomodulating activities. CAPE is the phenethyl alcohol ester of caffeic acid and a bioactive component of honeybee hive propolis. In addition, CAPE inhibits PDGF-induced proliferation of vascular smooth muscle cells through the activation of p38 mitogen-activated protein kinase (MAPK) and hypoxia-inducible factor (HIF)-1alpha and subsequent induction of heme oxygenase-1 (HO-1). Purity 99.93%
Catalog No. 50-287-9851 Supplier TARGETMOL CHEMICALS INC Supplier No. T6429200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Caffeic Acid Phenethyl Ester (Phenylethyl Caffeate) (CAPE) inhibits the activation of nuclear transcription factor NF-kappa B and may suppress p70S6K and Akt-driven signaling pathways, with antineoplastic, cytoprotective and immunomodulating activities. CAPE is the phenethyl alcohol ester of caffeic acid and a bioactive component of honeybee hive propolis. In addition, CAPE inhibits PDGF-induced proliferation of vascular smooth muscle cells through the activation of p38 mitogen-activated protein kinase (MAPK) and hypoxia-inducible factor (HIF)-1alpha and subsequent induction of heme oxygenase-1 (HO-1). Purity 99.93%