Learn More
Selleck Chemical LLC Caffeic Acid Phenethyl Ester
SDP
Catalog No. 5000004792
Encompass_Preferred
Caffeic acid phenethyl ester (CAPE Phenylethyl Caffeate) is a potent and specific inhibitor of NF- B activation and also displays antioxidant immunomodulatory and antiinflammatory activities
Catalog No. 50-000-04792 Supplier Selleck Chemical LLC Supplier No. S7414200MG
Caffeic acid phenethyl ester (CAPE Phenylethyl Caffeate) is a potent and specific inhibitor of NF- B activation and also displays antioxidant immunomodulatory and antiinflammatory activities