Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Caffeine
SCI_ED

Catalog No. S25215A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25215A 100 g Lab Grade
S25215 25 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25215A
Add to Cart
Add to Cart
Mitosis studies

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Color White
Density 1.23
Assay Percent Range 1
Packaging Lab Grade
Linear Formula C8H10N4O2
Quantity 100 g
Formula Weight 194.19
Grade Laboratory
Chemical Name or Material Caffeine
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.