Learn More
Enzo Life Sciences Caffeine (5g). CAS: 58-08-2
SDP
Supplier:  Enzo Life Sciences ALX550322G005
Encompass_Preferred
CNS stimulant. Blocks adenosine A1 and A2A receptors. cAMP phosphodiesterase inhibitor. Interferes with the uptake and storage of Ca2+ by the sarcoplasmic reticulum in skeletal muscle. Prevents apoptosis and cell cycle effects induced by various chemicals. Inhibits cellular DNA repair mechanisms. Anti-inflammatory. Alternative name: 1,3,7-Trimethylxanthine, 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione. Purity: ≥99% (HPLC). Appearance: White to colorless powder. Solubility: Soluble in chloroform, 100% alcohol or hot water. Long Term Storage: Ambient.
Catalog No. 50-200-9534