Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine, 98.5%, specified according to the req. of USP/BP

Catalog No. AC108165000
Change view
Click to view available options
Quantity:
10 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108165000 500 g Plastic bottle
AC108160100 10 g Glass Bottle
AC108160025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108165000 Supplier Thermo Scientific Chemicals Supplier No. 108165000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC. | CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Acidity 0.2mL of 0.01M NaOH max.
Melting Point 234.0°C to 239.0°C
Color White
Density 1.2300g/mL
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. (total impurities) (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 26,461
Merck Index 15,1639
Specific Gravity 1.23
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: 1g/66 mL alcohol,1g/50 mL acetone,freely soluble in pyrrole,soluble in ethyl acetate
Sublimation Point 178 °C
Residual Solvents meets the requirements
Formula Weight 194.19
Percent Purity 98.5%
Related Substances 0.5% max. (TLC)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Caffeine, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-362-1
RTECSNumber EV6475000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.