Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine Citrate, MP Biomedicals™

Catalog No. MP520481880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP520481880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP520481880 Supplier MP Biomedicals, Inc Supplier No. 0520481880
Add to Cart
Add to Cart

Caffeine Citrate, White Powder

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-22-7
Molecular Formula C14H18N4O9
Molecular Weight (g/mol) 386.32
MDL Number MFCD00044986
InChI Key RCQXSQPPHJPGOF-UHFFFAOYSA-N
Synonym Citrated caffeine, Caffeine, Citrated, Caffeina citrate
PubChem CID 6241
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 235.6°C°C to 236.2°C (literature)
Color White
Loss on Drying 0.0034
Quantity 100 g
Solubility Information Soluble in water
Physical Form Powder
RTECSNumber EV6495400
storageNote1 Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.