Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine, citrated

Catalog No. AC461721000
Click to view available options
Quantity:
25 g
100 g

Caffeine, citrated, Quantity: 100g, Melting Point: 159.0 deg.C to 161.0 deg.C, Molecular Weight: 386.31, CAS: 69-22-7, Formula Weight: 386.31, Health Hazard 1: GHS Signal Word: Warning, Health Hazard 2: GHS H Statement Harmful if swallowed. Causes serious eye irritation. | CAS: 69-22-7 | C14H18N4O9 | 386.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-22-7
Molecular Formula C14H18N4O9
Molecular Weight (g/mol) 386.32
MDL Number MFCD00044986
InChI Key RCQXSQPPHJPGOF-UHFFFAOYSA-N
Synonym caffeine citrate, caffeine, citrated, caffeina citrate, citrated caffein, caffeine citrate 1:1, caffeine citrated, unii-u26eo4675q, citric acid, compd. with caffeine 1:1, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, mixt. with 3,7-dihydro-1,3,7-trimethyl-1h-purine-2,6-dione, dsstox_cid_26938
PubChem CID 6241
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;1,3,7-trimethylpurine-2,6-dione
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 159.0°C to 161.0°C
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: slightly soluble in dmso and methanol
Formula Weight 386.31
Chemical Name or Material Caffeine, citrated
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.