Learn More
Alpco Ltd Caffeine ELISA
SDP
Supplier:  Alpco Ltd 12CAFN96U
Encompass
The Caffeine ELISA is intended for highly sensitive, quantitative determination of Caffeine in human saliva, plasma, serum, food extracts, drinks, water, and other biological fluids. For research use only. Not intended for use in diagnostic procedures. The Caffeine ELISA shows virtually no cross-reaction with Paraxanthine (0.6%).
Catalog No. NC2284547