Learn More
U.S. Pharmacopeia Caffeine Melting Point Standard, 58-08-2, MFCD00005758, 1g
Catalog No. NC9635138
Encompass
Molecular formula C8H10N4O2, Molecular weight 194.19, Melting Point 455 - 462.2 °F (235 - 239 °C), Boiling Point 352.4 °F (178 °C) (sublimes), Synonyms: 1,3,7-Trimethylxanthine
Catalog No. NC9635138 Supplier U.S. Pharmacopeia Supplier No. 1086006
Molecular formula C8H10N4O2, Molecular weight 194.19, Melting Point 455 - 462.2 °F (235 - 239 °C), Boiling Point 352.4 °F (178 °C) (sublimes), Synonyms: 1,3,7-Trimethylxanthine