Learn More
Medchemexpress LLC Caffeine orange | 1632296-13-9 | 95.4% | 365.16 g/mol | C20H15BF3N3 | 10 MG
SDP
Catalog No. 5000296985
Encompass_Preferred
Caffeine orange is an aqueous-phase, turn-on fluorescent probe selective for caffeine. It emits orange fluorescence upon excitation at 532 nm and is supplied as a solid for research use in fluorescence-based caffeine detection and sensing applications.
  • Selective turn-on fluorescence response to caffeine in aqueous samples.
  • High extinction coefficient and photostability for reliable signal.
  • Narrow emission bandwidth producing orange emission under 532 nm excitation.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance for stock solution preparation.
  • Store protected from light at -20°C; in solution, store at -80°C for up to 6 months.
Catalog No. 50-002-96985 Supplier Medchemexpress LLC Supplier No. HYD226410MG
Caffeine orange is an aqueous-phase, turn-on fluorescent probe selective for caffeine. It emits orange fluorescence upon excitation at 532 nm and is supplied as a solid for research use in fluorescence-based caffeine detection and sensing applications.
  • Selective turn-on fluorescence response to caffeine in aqueous samples.
  • High extinction coefficient and photostability for reliable signal.
  • Narrow emission bandwidth producing orange emission under 532 nm excitation.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance for stock solution preparation.
  • Store protected from light at -20°C; in solution, store at -80°C for up to 6 months.