Learn More
Sigma Aldrich Fine Chemicals Biosciences Caffeine powder ReagentPlu500G
Catalog No. 502899145
Encompass
Caffeine is a widely ingested pharmacologically active substance. It belongs to the class of methylxanthines. It is a naturally occurring alkaloid found in coffee cocoa and other plants. Caffeine might be associated with osteoporosis cardiovascular risk arrhythmia and gastrointestinal disturbances. Caffeine is used to treat orthostatic hypotension bronchopulmonary dysplasia and apnea of prematurity (AOP).
Catalog No. 50-289-9145 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C0750500G
Caffeine is a widely ingested pharmacologically active substance. It belongs to the class of methylxanthines. It is a naturally occurring alkaloid found in coffee cocoa and other plants. Caffeine might be associated with osteoporosis cardiovascular risk arrhythmia and gastrointestinal disturbances. Caffeine is used to treat orthostatic hypotension bronchopulmonary dysplasia and apnea of prematurity (AOP).