Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine (Powder/USP/FCC), Fisher Chemical

Catalog No. O1728500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O1728-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O1728-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 237.8°C
Color White
Density 1.23g/cm³
Boiling Point 177.8°C
Ignition Residue 0.1% max. (FCC/USP)
Melting Range 235.0°C to 237.5°C (FCC), 235.0°C to 239°C (USP)
Packaging Glass Bottle
Quantity 500 g
UN Number 1544
Residual Solvents Pass Test
Formula Weight 194.19
Percent Purity 98.5 to 101.0% (FCC)
Grade FCC/USP
Physical Form Solid
Chemical Name or Material Caffeine
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 6.1: Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.