Learn More
Waters Corp WATERS CORPORATION Caffeine Standard, 1.000 mg/mL, MS Grade Acetonitrile, 10 mL
Catalog No. 50201676 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a Caffeine Standard used for system performance qualification in liquid chromatography. It consists of 10mL of 1.000 mg/mL Caffeine ± 0.5% in MS Grade Acetonitrile, designed for reliable detection across UV, MS, and ELSD methods.
  • Designed for system performance qualification.
  • Ideal for liquid chromatography systems.
  • Features reversed phase separation mode.
  • Compatible with UV, MS, and ELSD detection methods.
  • Supplied as a liquid formulation.
  • Stable at room temperature storage.
  • Has a shelf life of 36 months.
  • Ships under ambient conditions.
  • Each package contains 1 unit.
  • Contains hazardous material which requires special freight handling, with additional charges potentially applying.
Catalog No. 50-201-676 Supplier Waters Corp Supplier No. 700003233
This product is a Caffeine Standard used for system performance qualification in liquid chromatography. It consists of 10mL of 1.000 mg/mL Caffeine ± 0.5% in MS Grade Acetonitrile, designed for reliable detection across UV, MS, and ELSD methods.
  • Designed for system performance qualification.
  • Ideal for liquid chromatography systems.
  • Features reversed phase separation mode.
  • Compatible with UV, MS, and ELSD detection methods.
  • Supplied as a liquid formulation.
  • Stable at room temperature storage.
  • Has a shelf life of 36 months.
  • Ships under ambient conditions.
  • Each package contains 1 unit.
  • Contains hazardous material which requires special freight handling, with additional charges potentially applying.