Learn More
Medchemexpress LLC Caffeoyl alcohol | 3598-26-3 | 166.17 | 5 MG
SDP
Catalog No. 5000415597
Encompass_Preferred
Caffeoyl alcohol is a monomer constituting Catechyl lignin. Catechyl lignin is a linear homopolymer of caffeyl alcohol, a natural source of carbon fiber and high-value chemicals. It has a high purity of 99.89% and is supplied as a solid, white to off-white in color.
  • Structure classification: Phenols, polyphenols
  • Initial source: Plants, leguminosae, Medicago sativa L.
  • For research use only
Catalog No. 50-004-15597 Supplier Medchemexpress LLC Supplier No. HYN103735MG
Caffeoyl alcohol is a monomer constituting Catechyl lignin. Catechyl lignin is a linear homopolymer of caffeyl alcohol, a natural source of carbon fiber and high-value chemicals. It has a high purity of 99.89% and is supplied as a solid, white to off-white in color.
  • Structure classification: Phenols, polyphenols
  • Initial source: Plants, leguminosae, Medicago sativa L.
  • For research use only